Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ankyrin 1 Antibody (S388A-10), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259315R
Description
Ankyrin 1 Monoclonal specifically detects Ankyrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Ankyrin 1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANK1 | |
Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R | |
100 μL | |
286 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S388A-10 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ANK, ANK-1, ankyrin 1, erythrocytic, ankyrin-1, Ankyrin-R, Erythrocyte ankyrin, SPH1, SPH2 | |
Mouse | |
Protein G purified | |
Primary | |
Detects 200kDa. Does not cross-react with Ankyrin-B or Ankyrin-G. | |
Store at 4C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction