Learn More
Ankyrin 1 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S388A-10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259315IR
Catalog No. NBP259315IR
Add to cart
Description
Description
Ankyrin 1 Monoclonal antibody specifically detects Ankyrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Ankyrin 1
S388A-10
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
ANK1
Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 200kDa. Does not cross-react with Ankyrin-B or Ankyrin-G.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ANK, ANK-1, ankyrin 1, erythrocytic, ankyrin-1, Ankyrin-R, Erythrocyte ankyrin, SPH1, SPH2
Mouse
IgG2b
100 ul
Primary
286
Human, Mouse, Rat