Learn More
Ankyrin 3 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S106-20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259310V
Catalog No. NBP259310V
Add to cart
Description
Description
Ankyrin 3 Monoclonal antibody specifically detects Ankyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Ankyrin 3
S106-20
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
ANK3
Fusion protein 1000 C-terminal amino acids of human Ankyrin G encompassing all of Ankyrin G with the exception of Ankyrin repeats
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects >200kDa.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier
Mouse
IgG1
100 ul
Primary
288
Human, Mouse, Rat