Learn More
Ankyrin Brain Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: S105-13, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259374R
Catalog No. NBP259374R
Add to cart
Description
Description
Ankyrin Brain Monoclonal antibody specifically detects Ankyrin Brain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Ankyrin Brain
S105-13
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
ANK2
Synthetic peptide amino acids 203-496 of human Ankyrin-B
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 200kDa. No cross reactivity against Ankyrin-G.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Ank2, Ankyrin 2, Ankyrin 2 brain, Ankyrin B, Ankyrin brain, Ankyrin nonerythroid, Brain Ankyrin
Mouse
IgG1
100 ul
Primary
287
Human, Mouse, Rat