Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin repeat domain 39 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen Ankyrin repeat domain 39
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179652
View Documents
Novus Biologicals
NBP179652
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

Ankyrin repeat domain 39 Polyclonal specifically detects Ankyrin repeat domain 39 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Ankyrin repeat domain 39
Polyclonal
Rabbit
PBS and 2% Sucrose with 0.09% Sodium Azide
ankyrin repeat domain 39, ankyrin repeat domain-containing protein 39, MGC41816
ANKRD39
IgG
Affinity Purified
20 kDa
Western Blot
Unconjugated
RUO
NP_057550
51239
Synthetic peptide directed towards the N terminal of human ANKRD39. Peptide sequence IWSAALNGDLGRVKHLIQKAEDPSQPDSAGYTALHYASRNGHYAVCQFLL.
Primary
Store at -20C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit