Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKZF1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550601

Catalog No. PA550601


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ANKZF1 protein.

Plays a role in the cellular response to hydrogen peroxide and in the maintenance of mitochondrial integrity under conditions of cellular stress (PubMed:28302725). Involved in the endoplasmic reticulum (ER)-associated degradation (ERAD) pathway. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANKZF1
Polyclonal
Unconjugated
ANKZF1
1300008P06Rik; 2810025E10Rik; ankyrin repeat and zinc finger domain containing 1; ankyrin repeat and zinc finger domain-containing protein 1; ankyrin repeat containing protein RGD1359242; ANKZF1; D1Ertd161e; RGD1359242; Zinc finger protein 744; ZNF744
Rabbit
Antigen affinity chromatography
RUO
55139
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9H8Y5
ANKZF1
Synthetic peptide corresponding to a region derived from internal residues of human ankyrin repeat and zinc finger domain containing 1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.