Learn More
Abnova™ ANLN 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00054443T01
Catalog No. 89-933-477
Add to cart
Description
Description
Specifications
Specifications
293T
BC034692.1
pCMV-ANLN full-length
100 μL
Human
ANLN 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 45.4kDa
Transient overexpression cell lysate was tested with anti-ANLN antibody (Mfr. No. H00054443-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot