Learn More
BioVision IncSupplier Diversity Partner Annexin V-Cy5 Apoptosis Kit: Annexin V-Cy5 Apoptosis Detection Kit: Simple One Step Assay to Detect Apoptosis within 10 min. Detection: Flow Cytometry or Fluorescence Microscopy DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K10325
Annexin V Apoptosis Detection Kit is based on the observation that soon after initiating apoptosis, cells translocate the membrane phosphatidyl-serine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can be easily detected by staining with a fluorescent conjugate of Annexin V, a protein that has a high affinity for PS. The one-step staining procedure takes only 10 minutes. Detection can be analyzed by flow cytometry or by fluorescence microscopy. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9460