Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A1 Mouse anti-Human, Clone: CPTC-ANXA1-1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Annexin A1
Clone CPTC-ANXA1-1
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunoprecipitation 2-4 ug/mg protein, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122902
View Documents
Novus Biologicals
NBP30771420UG
20 μg
Each for $138.37
Add to cart
 
NB122901
View Documents
Novus Biologicals
NBP30771410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

Annexin A1
0.2 mg/ml
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Purified
RUO
Human
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Recombinant full-length human Annexin A1 protein
Monoclonal
Protein A or G purified
CPTC-ANXA1-1
Western Blot 1-2 ug/ml, Immunoprecipitation 2-4 ug/mg protein, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Protein Kinase, Signal Transduction
10 mM PBS with 0.05% BSA
301
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit