Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Mouse anti-Human, DyLight 488, Clone: OTI3A8, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270174G
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
Annexin A1 | |
OTI3A8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
Affinity Purified, Conjugated | |
ANXA1 | |
Full length recombinant protein of human Annexin A1 (NP_000691) produced in HEK293T cells. | |
Immunogen affinity purified | |
Protein Kinase, Signal Transduction | |
Primary | |
301.0 |
Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title