Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A1, Mouse anti-Human,Mouse,Rat,Primate, Alexa Fluor 350, Clone: 2F1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223485AF350

Catalog No. NB005114


Add to cart

Description

Description

Annexin A1 Monoclonal specifically detects Annexin A1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Annexin A1
2F1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Conjugated
ANXA1
Partial recombinant human Annexin A1 (between residues 100-300) expressed in E. coli [UniProt P04083]
Ammonium sulfate precipitation
RUO
Store at 4°C in the dark.
Monoclonal
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Mouse
IgG1
0.1 mL
Protein Kinase, Signal Transduction
Primary
Human, Mouse, Rat, Primate
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit