Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Rabbit anti-Human, Clone: RM424, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30917725UL
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Annexin A1 | |
Monoclonal | |
Unconjugated | |
PBS, 50% Glycerol, 1% BSA | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
RM424 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:200 | |
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
A peptide corresponding to the N-terminus of human Annexin A1 | |
25 μg | |
Protein Kinase, Signal Transduction | |
301 | |
Store at -20°C. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction