Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A1 Rabbit anti-Human, Mouse, Clone: 7H46L26, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 702057

Encompass

Catalog No. 70-205-7


Add to cart

Description

Description

This antibody is predicted to react with Monkey, Dog and Rabbit Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Annexin A1 Recombinant Monoclonal antibody specifically detects Annexin A1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blot
Specifications

Specifications

Annexin A1
Immunocytochemistry, Immunofluorescence, Western Blot
7H46L26
Unconjugated
ANXA1
Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; Anx1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; Calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; phospholipase A2 inhibitory protein; RP11-71A24.1
Protein corresponding to Human ANXA1 (aa 33-346)
Protein A
RUO
Antibody
Monoclonal
Human, Mouse
Liquid
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P04083, P10107
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
16952, 301
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit