Learn More
Annexin A2 Goat anti-Annexin A2, Biotin, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP261464B
Catalog No. NBP261464B
Add to cart
Specifications
Specifications
Annexin A2
LYOPH
Western Blot 1ug/mL, Simple Western 50ug/mL, Immunohistochemistry 5-15ug/mL, Knockout Validated
Affinity Purified
ANXA2
E. coli-derived recombinant human Annexin A2, Met1-Asp339, Accession # P07355
Affinity purified
Store at 4C in the dark.
Polyclonal
Detects human, mouse, and rat Annexin A2 in Western blots. In Western blots, no cross-reactivity with recombinant human Annexin A1, A3, A4, A5, A6, A7, A8, A9, A10, A11, or A13 is observed.
Human, Mouse, Rat
Immunohistochemistry, KnockDown, Western Blot
Biotin
Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with 0.05% Sodium Azide
annexin A2, Annexin II, annexin-2, ANX2, ANX2L4LPC2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, Chromobindin-8, LIP2PAP-IV, Lipocortin II, LPC2DP36, p36, Placental anticoagulant protein IV, Protein I
Goat
IgG
0.1 ml
Primary
302
Reconstitute at 0.2 mg/mL in sterile PBS.