Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A3 Mouse anti-Human, Rat, PE/Cy5.5, Clone: OTI1A5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270179PECY55

Catalog No. N270179PEC5


Add to cart

Description

Description

Annexin A3 Monoclonal antibody specifically detects Annexin A3 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

Annexin A3
OTI1A5
Flow Cytometry
Affinity Purified, Conjugated
ANXA3
Full length recombinant protein of human Annexin A3 (NP_005130) produced in HEK293T cells.
Immunogen affinity purified
Signal Transduction
Primary
306.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
35-alpha calcimedin, annexin A3,1,2-cyclic-inositol-phosphate phosphodiesterase, Annexin III, annexin-3, calcimedin 35-alpha, Lipocortin III, PAP-III, Placental anticoagulant protein III, placental anticoagulant protein III, calcimedin 35-alpha
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit