Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A7 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1C3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270181AF647

Catalog No. N270181A647


Add to cart

Description

Description

Annexin A7 Monoclonal antibody specifically detects Annexin A7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Annexin A7
OTI1C3
Western Blot
Affinity Purified, Conjugated
ANXA7
Recombinant protein fragment corresponding to amino acids 105-411 of human Annexin A7 (NP_004025) produced in E.coli.
Immunogen affinity purified
Cancer
Primary
310.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
annexin A7, ANX7, ANXA7, SNX, SYNEXIN
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit