Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A7 Mouse anti-Human, DyLight 650, Clone: OTI1C3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270181C
Description
Annexin A7 Monoclonal antibody specifically detects Annexin A7 in Human samples. It is validated for Western Blot.Specifications
Annexin A7 | |
OTI1C3 | |
Western Blot | |
Affinity Purified, Conjugated | |
ANXA7 | |
Recombinant protein fragment corresponding to amino acids 105-411 of human Annexin A7 (NP_004025) produced in E.coli. | |
Immunogen affinity purified | |
Cancer | |
Primary | |
310.0 |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
annexin A7, ANX7, ANXA7, SNX, SYNEXIN | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title