Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kits

Catalog No. 50246345
Encompass_Preferred
Click to view available options
Quantity:
100 Tests
20 Tests
200 Test
200 Tests
300 Tests
50 Tests
Conjugate:
APC, Propidium Iodide
Biotin, Propidium Iodide
FITC, Propidium Iodide
PCP-eFluor 710
PE, 7-AAD
PE/Cy7
eFluor 450, 7-AAD
Excitation/Emission:
405, 546/450, 647
488, 535/520, 617
650, 535/660, 617
NA, 535/NA, 617

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.

Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.

Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

TRUSTED_SUSTAINABILITY

Specifications

Description eBioscience Annexin V Apoptosis Detection Set PE-Cyanine7
Quantity 50 Tests
Kit Contents Contains:
• Annexin V Binding Buffer (10X), 1 bottle (30 mL)
• Annexin V PE-Cyanine7, 1 vial (0.25 mL)

Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Product Type Apoptosis Detection Kit
Content And Storage Handling Conditions:
Store at 2-8°C. Avoid temperature fluctuations.
Shipping Condition Wet Ice
Conjugate PE/Cy7
For Use With (Equipment) Flow Cytometer
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.