Learn More
BioVision IncSupplier Diversity Partner Annexin V-PE-Cy5 Apoptosis Detection Kit: Annexin V-PE-Cy5 Apoptosis Detection Kit: Simple One Step Assay to Detect Apoptosis within 10 min. No need to fix the cells. Detection: Flow Cytometry or Fluorescence Microscopy DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K12925
The Annexin V-PE-Cy5 Apoptosis Detection Kit is based on the observation that soon after initiating apoptosis, most cell types translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can be easily detected by staining with a fluorescent conjugate of Annexin V, a protein that has a strong natural affinity for PS. The one-step staining procedure takes only 10 minutes. In addition, the assay can be directly performed on live cells, without the need for fixation. Results can be analyzed by flow cytometry or fluorescence microscopy. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9543