Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin V Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
$236.88 - $608.00
Specifications
Antigen | Annexin V |
---|---|
Concentration | 0.5 mg/mL |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001930
|
Novus Biologicals
NB1001930 |
0.05 mg |
Each for $378.00
|
|
NB100193001
|
Novus Biologicals
NB10019300.1MG |
0.1 mg |
Each for $608.00
|
|
NB017428
|
Novus Biologicals
NB10019300.025MG |
0.025 mg |
Each for $236.88
|
|
Description
Annexin V Polyclonal specifically detects Annexin V in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Annexin V | |
Polyclonal | |
Rabbit | |
P08758 | |
308 | |
IgG | |
Affinity Purified | |
36 kDa |
0.5 mg/mL | |
Unconjugated | |
Human, Mouse, Rat | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
ANXA5 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title