Learn More
ANO1 Mouse anti-Human, Clone: OTI1H2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803368
Catalog No. 50-166-6454
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
Specifications
Specifications
ANO1
Monoclonal
Unconjugated
ANO1
Lyophilized
ANO1
Human recombit protein fragment corresponding to amino acids 1-333 of human ANO1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI1H2
PBS with 8% trehalose and no preservative; pH 7.3
Q5XXA6
DOG1, ORAOV2, TAOS2, TMEM16A
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55107