Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 219011AP

Catalog No. 50-173-3629


Add to cart

Description

Description

TMEM16K is a multi-pass membrane protein. It belongs to the anoctamin family. TMEM16K may act as a calcium-activated chloride channel.
Specifications

Specifications

ANO10
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BH79, Q9NW15
ANO10, anoctamin 10, TMEM16K, Transmembrane protein 16K
Rabbit
IgG
150 μL
-20°C
Primary
102566, 55129
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ANO10
Liquid
ANO10
ANO10 Fusion Protein Ag13983
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit