Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANO2 Antibody (CL10631), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32117825UL
Description
ANO2 Monoclonal antibody specifically detects ANO2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ANO2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CL10631 | |
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
anoctamin 2, anoctamin-2, DKFZp434P102, TMEM16B, transmembrane protein 16B (eight membrane-spanning domains) | |
This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q9NQ90. Actual immunogen sequence is proprietary. | |
25 μg | |
Neuroscience | |
57101 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction