Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANO9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569330
Description
This target displays homology in the following species: Human: 100%.
Does not exhibit calcium-activated chloride channel (CaCC) activity. Can inhibit the activity of ANO1. Jikuya H., DNA Res. 10:49-57(2003). The MGC Project Team, Genome Res. 14:2121-2127(2004).
Specifications
ANO9 | |
Polyclonal | |
Unconjugated | |
ANO9 | |
5430425C04Rik; ANO9; anoctamin 9; anoctamin-9; p53-induced gene 5 protein; PIG5; RGD1560565; TM16J; TMEM16J; TP53I5; transformation related protein 53 inducible protein 5; transmembrane protein 16J; Trp53 inducible protein 5; Trp53i5; tumor protein p53 inducible protein 5; tumor protein p53-inducible protein 5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
338440 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A1A5B4 | |
ANO9 | |
synthetic peptide directed towards the middle region of human ANO9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction