Learn More
Advanced Instruments Anoxomat™ System Ergonomic Anaerobic Culture Jars
Advanced Instruments Anoxomat™ System Ergonomic Anaerobic Culture Jars maximize incubator, and laboratory space
$131.72 - $549.00
Specifications
For Use With (Application) Anoxomat™ Ergonomic Anaerobic Culture Jars
For Use With (Equipment) Anoxomat™ Anaerobic Jar System
View More Specs
Products 4
Catalog Number Mfr. No. Item Description Price Quantity & Availability  
Catalog Number Mfr. No. Item Description Price Quantity & Availability  
22-047-542
View Documents
Advanced Instruments
AN2033
Lid
Each for $219.85
Add to cart
22-047-606
View Documents
Advanced Instruments
AN2040
Small Jar
Each for $131.72
Add to cart
22-047-604
View Documents
Advanced Instruments
AJ9049
Small Ergonomic
Each for $396.00
Add to cart
22-047-605
View Documents
Advanced Instruments
AJ9050
Large Ergonomic
Each for $549.00
Add to cart
Description
Description
  • Stacking
    • Nested stacking, enabling more jars to fit easily into incubators
  • Lid System
    • Unique lid clamp-on system creates a secure seal with 1-simple click
  • Lid Coupling
    • Recessed, snap-shut lid coupling easily connects to gas supply
  • Grip Handle
    • Fold-down grip handle for safe, and easy transport
  • Construction
    • Lightweight construction, and compact design
Specifications
Specifications
Anoxomat™ Ergonomic Anaerobic Culture Jars
Anoxomat™ Anaerobic Jar System