Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advanced Instruments Anoxomat™ System Standard Anaerobic Culture Jars

Advanced Instruments Anoxomat™ System Standard Anaerobic Culture Jars guarantee reliable and reproducible growth

$123.34 - $917.00

Specifications

For Use With (Equipment) Anoxomat™ Anaerobic Jar System
Storage Requirements 50 to 91°F
View More Specs

Products 9
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
22-047-537
View Documents
Advanced Instruments
AAM0040060
Jar Only - Small Standard
Each for $444.28
Add to cart
 
22-047-541
View Documents
Advanced Instruments
AAM0090180
Small Standard - Jar Clamp Assembly
Each for $123.34
Add to cart
 
22-047-602
View Documents
Advanced Instruments
AJ9025
Small Standard Jar
Each for $634.07
Add to cart
 
22-047-536
View Documents
Advanced Instruments
AAM0040055
Jar Only - Large Standard
Each for $595.60
Add to cart
 
22-047-540
View Documents
Advanced Instruments
AAM0090060
Large Standard - Jar Lid Assembly
Each for $131.43
Add to cart
 
22-047-603
View Documents
Advanced Instruments
AJ9028
Large Standard Jar
Each for $917.00
Add to cart
 
22-047-624
View Documents
Advanced Instruments
AN3155
Standard Jars - Small and Large
Each for $377.26
Add to cart
 
22-047-538
View Documents
Advanced Instruments
AAM0040070
Small Standard - Jar Lid Assembly This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
22-047-535
View Documents
Advanced Instruments
AAM0040040
Lid Only - Large Standard This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Lid
    • Strong lid clamp to meet microbiology standards, clear lid for plate visibility
  • Connection
    • Quick-connect coupling for easy connection
  • Plates
    • Microtiter plates, 16 mm tubes, and 50-150 mm plates compatible
  • Guaranteed Proof
    • Guaranteed anaerobic, microaerophilic, capnophillic and hypoxic growth
Specifications

Specifications

Anoxomat™ Anaerobic Jar System
50 to 91°F
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit