Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD39 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548134

Catalog No. PA548134

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ANKRD39
Polyclonal
PBS with 0.09% sodium azide
Q53RE8, Q9D2X0
Ankyrin repeat domain-containing protein 39, ANKRD39
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
109346, 51239
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANKRD39
Liquid
ANKRD39
KLH conjugated synthetic peptide between 137-165 amino acids from the C-terminal region of human ANR39.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit