Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD46 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548370

Catalog No. PA548370

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

ANKRD46 encodes a protein containing multiple ankyrin repeats. Ankyrin domains function in protein-protein interactions in a variety of cellular processes. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

ANKRD46
Polyclonal
PBS with 0.09% sodium azide
Q86W74
Ankyrin repeat domain-containing protein 46, Ankyrin repeat small protein, ANK-S, ANKRD46
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
157567
Western Blot
Unconjugated
ANKRD46
Liquid
ANKRD46
KLH conjugated synthetic peptide between 17-46 amino acids from the N-terminal region of human ANR46.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit