Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ 73-500 Touch N Tuff™ Sensitive Neoprene Gloves

Provides chemical splash resistance against wide variety of chemicals

Manufacturer:  Ansell™ 320006

 View more versions of this product

Catalog No. 19-150-186


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Long, beaded
Ideal for double donning and dry applications
Neoprene
4.3 in.
5 mil
Powdered
Sterile
305mm
Anatomic
Yes
8.5
110mm
CE, ISO 9001, ISO 13485 and ISO 14001
Cream
12 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit