Ansell™ AlphaTEC™ Gloves
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals
Manufacturer: Ansell™ 58530 8 219002
 View more versions of this product
Catalog No. 19-161-446
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Glove Cuff Style Gauntlet
    For Use With (Application) Chemical plants and transportation, chemical processing; oil refining and petrochemicals; cleaning and de-greasing; exposure to chemicals, solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
    Length (Metric) 30.5cm
    Material Nitrile
    Size 8
    Thickness 13mil
    Length (English) 12 in.
    Lining Knit Acrylic
    Color Red with black coating
  • Description & Specifications
Related Products
Product Content Feedback