Ansell™ AlphaTEC™ Gloves
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals
Manufacturer: Ansell™ 58530 10 219004
 View more versions of this product
Catalog No. 19-161-448
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Glove Cuff Style Gauntlet
    For Use With (Application) Chemical plants and transportation, chemical processing; oil refining and petrochemicals; cleaning and de-greasing; exposure to chemicals, solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
    Material Nitrile
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Lining Knit Acrylic
    Size 10
    Thickness 13mil
    Color Red with black coating
  • Description & Specifications
Related Products
Product Content Feedback