Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 58-535B Series Nitrile Immersion Gloves

Catalog Number 19161450
Click to view available options
Size:
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.

Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals
  • Chemical resistant
  • Material fully coated nitrile gloves
  • Thickness: 11 mil
  • Embossed grip surface for superior wet and oil grip
  • Red with black coating
  • Comfortable design reduces fatigue
The patented Ansell Grip Technology™ ensures that the coating polymer does not penetrate the knitted liner during manufacture. The coating is therefore consistent and sound, significantly reducing the likelihood of chemical leakage onto the skin. Available in two lengths 305mm and 355mm.
Certifications/Compliance FDA 21 CFR 177-2600
Color Green, Black
Latex Free Yes
Material Nitrile
Size 9
Thickness (English) 13 mil
Thickness (Metric) 0.33 mm
For Use With (Application) Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Liner Material Knit Lined Acrylic
Series 58-535
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit