Ansell™ Barrier™ Gloves
Provides excellent chemical resistance to most type of chemicals including warfare agents and biological hazards. Ansell™ Barrier™ Gloves include welded-on plastic ring for mounting in the Trellchem™ Bayonet ring system.
Manufacturer: Ansell™ 2100 10 902213
 View more versions of this product
Catalog No. 19-034-564E
  Check Availability
Add to Cart
    Specifications
    Certifications ANSI EN 374
    Hand Hand-specific
    For Use With (Application) aircraft assembly, chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, graphic arts and printing cleanup, handling Acids, Alkalies and Salts, handling Ketones, handling specialty chemicals, handling strong o
    Length (Metric) 38 to 41cm
    Material Five Layer Laminated Film
    Size 10
    Thickness 2.5mil
    Color White
    Length (English) 14.9 to 16.14 in.
    Lining Non-woven
    • Provide flexibility and tactility
    • Made from an exceptionally thin chemical-resistant, five-layer laminated film
    • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
    • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
    • Hand specific
    • Broad spectrum chemical resistance
    • Wing-thumb for added wearer comfort
  • Description & Specifications
Related Products
Product Content Feedback