Learn More
Ansell™ Barrier™ Gloves
Provides excellent chemical resistance to most type of chemicals including warfare agents and biological hazards. Ansell™ Barrier™ Gloves include welded-on plastic ring for mounting in the Trellchem™ Bayonet ring system.
Manufacturer:  Ansell™ 2100 11 902214
 View more versions of this product
Catalog No. 19-034-564F
  Check Availability
Add to cart
Specifications
Description
  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort
Description & Specifications
Specifications
Five Layer Laminated Film
aircraft assembly, chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, graphic arts and printing cleanup, handling Acids, Alkalies and Salts, handling Ketones, handling specialty chemicals, handling strong o
Laminated Film
2.5mil
White
38 to 41cm
Hand-specific
Non-woven
11
ANSI EN 374
14.9 to 16.14 in.