Ansell™ Barrier™ Gloves
Provides excellent chemical resistance to most type of chemicals including warfare agents and biological hazards. Ansell™ Barrier™ Gloves include welded-on plastic ring for mounting in the Trellchem™ Bayonet ring system.
Manufacturer: Ansell™ 2100 6 902209
 View more versions of this product
Catalog No. 19-034-564A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Certifications ANSI EN 374
    Hand Hand-specific
    For Use With (Application) aircraft assembly, chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, graphic arts and printing cleanup, handling Acids, Alkalies and Salts, handling Ketones, handling specialty chemicals, handling strong o
    Length (English) 14.9 to 16.14 in.
    Length (Metric) 38 to 41cm
    Lining Non-woven
    Material Five Layer Laminated Film
    Size 6
    Thickness 2.5mil
    Color White
    • Provide flexibility and tactility
    • Made from an exceptionally thin chemical-resistant, five-layer laminated film
    • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
    • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
    • Hand specific
    • Broad spectrum chemical resistance
    • Wing-thumb for added wearer comfort
  • Description & Specifications
Related Products
Product Content Feedback