Ansell™ Chemical- and Flame-Resistant Bib Pants

Lightweight and breathable CPC garments made with Gore™ ChemSplash™ fabric reduce overheating and fatigue while preventing liquid chemical splash penetration

Manufacturer: Ansell™ 66 662 2X966412

 View more versions of this product

Catalog No. 17-987-449F

  Check Availability
Add to Cart

Alert

Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.


    Specifications

    Material CPC Polyester
    Size 2X-Large
    Size (English) Waist 60 in.
    Certifications/Compliance NFPA, ASTM F903
    Suspender Elastic
    For Use With (Application) Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals
    Closure Type Hook and loop, double front flap
    Length (English) Inseam 31 in.
    Length (Metric) Inseam 79cm
    Color Red
    Construction Trilaminate

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit