Ansell™ CPP Vinyl Aprons
Heavy-duty protection in a variety of styles
Manufacturer: Ansell™ 56 101 50950103
 View more versions of this product
Catalog No. 19-058744
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Color White
    Size 35 x 50
    Material Vinyl
    Highest-quality virgin vinyl in 4, 6, 8, 12 and 18mil thicknesses
    Benefits:
    • Superior resistance to chemicals, fats, oils, greases, punctures and abrasions
  • Optional features:
    • Stomach patches for extended wear
    • Raw or sewn edges
    • Three-piece ties
    • Rough texture
    Food Preparation, Processing and Handling:
    • Beef/pork/poultry/fish processing
    • Dairy processing
    Public Utilities:
    • Cleaning and de-greasing
    • Industrial maintenance
    • Janitorial and maintenance work
    Chemical:
    • Chemical processing, oil refining and petrochemicals
  • Description & Specifications
Related Products
Product Content Feedback