Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Crusader™ Flex Gloves

Non-woven felt insulation and a full-length heat barrier designed for intermittent handling of objects up to 400°F. Ansell™ Crusader™ Flex Gloves can be used in industries such as food service, and glass and metal fabrication.

Manufacturer:  Ansell™ 42325 9 209307

 View more versions of this product

Catalog No. 19-019842



  Check Availability
Add to cart

Specifications

Description

  • Front-and-back heat-insulated styles
  • Provides protection from radiant heat and intermittent back-of-hand contact with hot objects
  • Provides excellent grip when handling dry or oily surfaces
  • Gauntlet slip-on cuff
  • Contains no asbestos
  • Length: 10 in. (25.4cm)

 

Not a replacement for silicate gloves. Designed for intermittent handling of objects up to 400°F (204°C).

Description & Specifications

Specifications

Fully coated
Launderable
Nitrile
Food service, glass and metal fabrication
9,Medium
10 in.
ANSI 2
Full-length heat barrier
Gauntlet Slip-on
Non-woven felt
Light Gray
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit