Ansell™ Edge™ Gloves

Outperformed 8 oz. cotton gloves ten to one

Manufacturer: Ansell™ 40400 8 218711

 View more versions of this product

Catalog No. 19-019825

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Cleaning Procedure Machine washable
    Exterior Finish Smooth
    Glove Cuff Style Knit
    For Use With (Application) fabrication, assembly, maintenance and shipping and receiving
    Coated Area Palm, finger
    Color Gray, white
    Material Nitrile
    Size 8

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit