Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Encore Acclaim Latex Sterile Gloves

Latex, powder-free surgical glove with a comfortable, natural fit and feel.

Manufacturer:  Ansell™ 5795007

Catalog No. 19-166-682


Add to cart

Description

Description

Featuring Ansell formers and glove formulation, this glove's combination of fit and comfort together with optimized tensile strength, puncture and tear resistance properties enables healthcare workers to perform at their very best.

  • Proprietary formulation delivers a soft and sensitive yet robust surgical glove
  • Unique anti-slip, smooth external surface treatment for exceptional instrument handling and grip
  • DERMASHIELD Technology™ internal coating facilitates dry and damp donning as well as intra-operative changes
  • Tested for use with chemotherapy drugs
Specifications

Specifications

Smooth
Yes
Hand-specific
9
11.5cm
Powder-free
Sterile
29.7cm
11.7 x 4.5 in. (29.7 x 11.5cm)
Beaded/Rolled
Rubber, natural
4.5 in.
9mil
Cream
11.7 in.
ASTM D 1076, ISO9001, ISO13485, ASTM D 3577, Type I
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit