Ansell™ FL200 Flock-Lined Latex Gloves
Superior tensile strength and sensitivity
Manufacturer: Ansell™ 00254 7 192541
 View more versions of this product
Catalog No. 19-014489
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Exterior Finish Pebble embossed
    Glove Cuff Style Pinked
    For Use With (Application) chemical plants, janitorial services, automotive assembly, plant maintenance, agriculture, Petroleum refineries, canning and pharmaceutical manufacturing, resist Acids, Detergents, Salts, Caustics, animal fats and Alcohols
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Lining Flock
    Material Natural Rubber
    Powdered No
    Size 7
    Sterile No
    Thickness 20mil
    Color Sky blue
  • Description & Specifications
Related Products
Product Content Feedback