Learn More
Ansell™ Hycar™ Nitrile/PVC Blend Aprons
Offer better resistance to hydrocarbon solvents than neoprene
Manufacturer:  Ansell™ YH 45HIB
 View more versions of this product
Catalog No. 17-987-108B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Good for use with animal fats and greases, especially in cold conditions.
Description & Specifications
Specifications
45 in.
35 in.
Yellow
Nitrile,PVC
18 oz.
114.3cm
88.9cm
Heavy Weight
45 x 35 in. (114.3 x 88.9cm)