Ansell™ HyFlex™ CR Gloves

For a sure grip in light oil applications; hides soil

Manufacturer: Ansell™ 11500 11 205548

 View more versions of this product

Catalog No. 19-152-148

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    Hem Color Blue
    For Use With (Application) handling and assembly of small, sharp, and/or pointed parts, handling wire, sheet Metal assembly
    Material Kevlar™
    Size 11
    Weight Medium weight
    Coating Material Foam Nitrile
    Color Yellow, black
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit