Ansell™ HyFlex™ CR2 Gloves with Dyneema™
Offer excellent abrasion resistance, cut protection, dexterity and comfort
Manufacturer: Ansell™ 11627 6 205686
 View more versions of this product
Catalog No. 19-168-50
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Knit
    Hem Color Purple
    For Use With (Application) Plastics injection and molding, Glass handling, sheet Metal assembly (even when slightly Oily), handling sharp, small parts
    Material Dyneema™, Lycra™
    Size 6
    Weight Medium weight
    Coating Material Polyurethane
    Color Gray
    Cut Resistance Rating Level 2
  • Description & Specifications
Related Products
Product Content Feedback