Learn More
Ansell™ HyFlex™ CR2 Gloves with Dyneema™
Offer excellent abrasion resistance, cut protection, dexterity and comfort
Manufacturer:  Ansell™ 205709
 View more versions of this product
Catalog No. 19-168-55
Add to cart
Description
Description
Polyurethane dipped; Dyneema and Lycra™ liner.
  • Palm coating provides dry grip and longer wear
  • Natural fit liner reduces hand fatigue
  • Excellent grip in dry environments
  • Effective control in minimally oily environments
  • Breathable liner increases worker acceptance and safety compliance
  • Machine launderable
Compliance
ANSI™ Level 2 cut protection.
Specifications
Specifications
Polyurethane
Knit
Plastics injection and molding, Glass handling, sheet Metal assembly (even when slightly Oily), handling sharp, small parts
11
Gray
Level 2
Blue
Dyneema™, Lycra™
Medium weight