Learn More
Ansell™ HyFlex™ Cut-Resistant Gloves
Offer a combination of comfort, softness, coolness and cut protection
Manufacturer:  Ansell™ 288738
 View more versions of this product
Catalog No. 19-168-464
Add to cart
Description
Description
  • Constructed of DSM Dyneema™, Spandex™ and nylon
  • Ergnomic design promotes wearer comfort
  • Proprietary black polyurethane coating provides sure grip
  • Can be washed and reused without any loss of cut protection
Compliance
ANSI™ Level 2 cut protection.
Specifications
Specifications
Polyurethane
Knit
Glass manufacturing, assembly of white goods, Plastics injection and moulding, automotive, assembling Metal parts and components, handling of sharp edged objects, preassembling, cutting of small dry or lightly Oiled parts and maintenance
10
Black, White
Level 2
Black
Dyneema™, Lycra™
Light weight