Ansell HyFlex Fully Dipped Light Duty Gloves with INTERCEPT™ and GRIP Technology
Ultralight weight 18 ga cut resistant glove with better grip performance
Manufacturer: Ansell 11539 8
 View more versions of this product
Catalog No. 19-010-132
  Check Availability
Add to Cart
    Specifications
    Cuff Style Knitwrist
    Description Fully dipped finishing
    For Use With (Application) Assembly of small parts, Light duties, Handling smooth, slippery or sharp-edged materials like glass, sheet metal, tile and veneer, Maintenance
    Material INTERCEPT™ Yarn; Nitrile
    Cut Resistance Rating Level 2
    Size 8
    Style 11-539
    Thickness 18 ga
    Coating Material Nitrile
    Product Type Ultralight weight cut resistant glove
    Length (Metric) 220-270mm
    Liner Material Ansell INTERCEPT™ Technology
    Color Anthracite
  • Description & Specifications
Related Products
Product Content Feedback