Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-600W White Nylon Gloves with Polyurethane Coating

Polyurethane coating on the palm improves tactile sensitivity, while the white color palette is useful for clean applications.

$83.68

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A1
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-152-157
View Documents
Ansell
205566
7
Dozen of 12 for $83.68
Case of 12 DZ for $955.88
Add to cart
 
19-152-158
View Documents
Ansell
205567
8
Dozen of 12 for $83.68
Case of 12 DZ for $955.88
Add to cart
 
Description

Description

  • Designed for light & delicate applications in dry environments
  • Bare-handed dexterity with protection for light duty applications
  • Polyurethane coating on the palm enhances tactile sensitivity
  • White color is ideal for clean applications
Specifications

Specifications

ANSI 3
White
Automotive, Machinery/Equipment, and Metal Fabrication
Nylon
15
Polyurethane
ANSI A1
Knit
11-600W
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit