Learn More
Ansell™ HyFlex™ Palm Dipped Medium Duty, 13 Gauge Gloves w/INTERCEPT™ Technology
Offers better flexibility and fit with optimal cut protection
Manufacturer:  Ansell 11724 8 163832
 View more versions of this product
Catalog No. 19-010-102
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyurethane
Knitwrist
8
Gray,White
Level 3
Assembling metal parts and components, Light metal presses, Maintenance, Plastics injection and moulding, Primary assembly
11-724
208-261mm