Ansell™ Latex Canners and Handlers Gloves
Unlined industrial latex gloves
Manufacturer: Ansell™ 88392 9 193925
 View more versions of this product
Catalog No. 11-391-135C
  Check Availability
Add to Cart
Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
    Specifications
    Certifications 21 CFR 177-2600, FDA
    Exterior Finish Pebble embossed
    Glove Cuff Style Pinked
    For Use With (Application) canning and food processing, small parts assembly, pharmaceutical manufacturing and plant maintenance, resist Acids, Detergents, vegetable Oil, Salts, Caustics and Alcohols
    Length (Metric) 30.5cm
    Material Natural Rubber
    Powdered No
    Size 9
    Sterile No
    Color Natural
    Length (English) 12 in.
    Thickness 20mil
  • Description & Specifications
Related Products
Product Content Feedback